accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3

CAS RN 10599-69-6

IUPAC Name: 1-(5-methylfuran-2-yl)propan-1-one
Synonyms: 1-(5-methylfuran-2-yl)propan-1-one
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: BXLPZYAVKVFXEO-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results