accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10599-70-9

IUPAC Name: 1-(2,5-dimethylfuran-3-yl)ethan-1-one
Synonyms: 1-(2,5-dimethylfuran-3-yl)ethanone
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: KBSVBCHYXYXDAG-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)OC(C)=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results