accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-02-5

IUPAC Name: 1-oxacyclohexadecan-2-one
Synonyms: exaltolide cyclopentadecanolide pentadecalactone
Molecular Weight (g/mol): 240.39
Molecular Formula: C15H28O2
InChi Key: FKUPPRZPSYCDRS-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCCCCO1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (4)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)