accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 106-18-3

IUPAC Name: butyl dodecanoate
Synonyms: dodecanoic acid, butyl ester
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: NDKYEUQMPZIGFN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCCCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results