accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 106-31-0

IUPAC Name: butanoyl butanoate
Synonyms: butyric anhydride
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: YHASWHZGWUONAO-UHFFFAOYSA-N
SMILES: CCCC(=O)OC(=O)CCC
Molecular Weight (g/mol) 
  • (3)
  • (8)
Percent Purity 
  • (1)
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (3)
  • (8)
  • (1)
  • (2)
  • (8)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (5)