accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 106-32-1

IUPAC Name: ethyl octanoate
Synonyms: ethyl octanoate ethyl octanoate (natural)
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: YYZUSRORWSJGET-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)OCC
Boiling Point 
  • (5)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)