accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 106-38-7

IUPAC Name: 1-bromo-4-methylbenzene
Synonyms: P-bromotoluene
Molecular Weight (g/mol): 171.04
Molecular Formula: C7H7Br
InChi Key: ZBTMRBYMKUEVEU-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (8)
Percent Purity 
  • (3)
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (8)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)