CAS RN 106-40-1
4-Bromoaniline 99.0+%, TCI America™
CAS: 106-40-1 Molecular Formula: C6H6BrN Molecular Weight (g/mol): 172.025 MDL Number: MFCD00007822 InChI Key: WDFQBORIUYODSI-UHFFFAOYSA-N Synonym: p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine PubChem CID: 7807 IUPAC Name: 4-bromoaniline SMILES: C1=CC(=CC=C1N)Br
4-Bromoaniline, 99+%
CAS: 106-40-1 Molecular Formula: C6H6BrN Molecular Weight (g/mol): 172.02 InChI Key: WDFQBORIUYODSI-UHFFFAOYSA-N Synonym: p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine PubChem CID: 7807 IUPAC Name: 4-bromoaniline SMILES: C1=CC(=CC=C1N)Br
4-Bromoaniline, 96%, pract.
CAS: 106-40-1 Molecular Formula: C6H6BrN Molecular Weight (g/mol): 172.02 MDL Number: MFCD00007822 InChI Key: WDFQBORIUYODSI-UHFFFAOYSA-N Synonym: p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine PubChem CID: 7807 IUPAC Name: 4-bromoaniline SMILES: C1=CC(=CC=C1N)Br
4-Bromoaniline, 98+%
CAS: 106-40-1 Molecular Formula: C6H6BrN Molecular Weight (g/mol): 172.025 MDL Number: MFCD00007822 InChI Key: WDFQBORIUYODSI-UHFFFAOYSA-N Synonym: p-bromoaniline,4-bromobenzenamine,benzenamine, 4-bromo,p-bromophenylamine,aniline, p-bromo,4-bromo-aniline,4-bromobenzeneamine,p-bromo aniline,4-bromanilinu,4-bromo-phenylamine PubChem CID: 7807 IUPAC Name: 4-bromoaniline SMILES: C1=CC(=CC=C1N)Br
Sigma Aldrich 4-Bromoaniline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 4-Bromoaniline
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.