CAS RN 106-43-4
4-Chlorotoluene, 98%
CAS: 106-43-4 Molecular Formula: C7H7Cl Molecular Weight (g/mol): 126.58 MDL Number: MFCD00000631 InChI Key: NPDACUSDTOMAMK-UHFFFAOYSA-N Synonym: 4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n PubChem CID: 7810 ChEBI: CHEBI:34401 IUPAC Name: 1-chloro-4-methylbenzene SMILES: CC1=CC=C(Cl)C=C1
4-chlorotoluene, 98%, Thermo Scientific Chemicals
CAS: 106-43-4 Molecular Formula: C7H7Cl Molecular Weight (g/mol): 126.58 MDL Number: MFCD00000631 InChI Key: NPDACUSDTOMAMK-UHFFFAOYSA-N Synonym: 4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n PubChem CID: 7810 ChEBI: CHEBI:34401 IUPAC Name: 1-chloro-4-methylbenzene SMILES: CC1=CC=C(Cl)C=C1
4-Chlorotoluene 98.0+%, TCI America™
CAS: 106-43-4 Molecular Formula: C7H7Cl Molecular Weight (g/mol): 126.58 MDL Number: MFCD00000631 InChI Key: NPDACUSDTOMAMK-UHFFFAOYSA-N Synonym: 4-chlorotoluene,p-chlorotoluene,benzene, 1-chloro-4-methyl,p-tolyl chloride,toluene, p-chloro,1-methyl-4-chlorobenzene,4-chloro-1-methylbenzene,monochlorotoluene,para-chlorotoluene,unii-q8r236h42n PubChem CID: 7810 ChEBI: CHEBI:34401 IUPAC Name: 1-chloro-4-methylbenzene SMILES: CC1=CC=C(Cl)C=C1
Sigma Aldrich 4-Chlorotoluene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
4-Chlorotoluene, SPEX CertiPrep™
CAS: 106-43-4 Molecular Formula: C7H7Cl Molecular Weight (g/mol): 126.58 MDL Number: MFCD00000631 InChI Key: NPDACUSDTOMAMK-UHFFFAOYSA-N PubChem CID: 7810 ChEBI: CHEBI:34401 IUPAC Name: 1-chloro-4-methylbenzene SMILES: CC1=CC=C(Cl)C=C1