accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-48-9

IUPAC Name: 4-chlorophenol
Synonyms: chlorophenol P-chlorophenol 4-chlorophenol ion (1+) parachlorophenol preparation sodium salt 4-chlorophenol parachlorophenol (substance) ccris 642 applied 3-78 parachlorophenol potassium salt 4-chlorophenol
Molecular Weight (g/mol): 128.56
Molecular Formula: C6H5ClO
InChi Key: WXNZTHHGJRFXKQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (7)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (4)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (3)
  • (2)
  • (7)
  • (6)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)