accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-50-3

IUPAC Name: benzene-1,4-diamine
Synonyms: p-phenylenediamine pPDA P-phenylenediamine paraphenylene-diamine PPDA p-phenylenediamine base
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: CBCKQZAAMUWICA-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (7)
  • (9)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (4)
  • (1)
  • (2)
  • (6)
  • (7)
  • (9)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)