accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOHN

CAS RN 106-57-0

IUPAC Name: piperazine-2,5-dione
Synonyms: 2,5-piperazinedione diglycolyl diamide glycine, bimol. cyclic peptide cycloglycylglycine α,gamma-diacipiperazine cyclodiglycine bimol. cyclic peptide glycine cyclo(glycylglycyl) glycylglycine lactam
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: BXRNXXXXHLBUKK-UHFFFAOYSA-N
SMILES: O=C1CNC(=O)CN1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (4)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (6)