accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOCH2

CAS RN 106-63-8

IUPAC Name: 2-methylpropyl prop-2-enoate
Synonyms: isobutyl acrylate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: CFVWNXQPGQOHRJ-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)C=C
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)