accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 106-68-3

IUPAC Name: octan-3-one
Synonyms: 3-octanone
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: RHLVCLIPMVJYKS-UHFFFAOYSA-N
SMILES: CCCCCC(=O)CC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)