accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 106-73-0

IUPAC Name: methyl heptanoate
Synonyms: methyl heptanoate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: XNCNNDVCAUWAIT-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)OC
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results