accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-92-3

IUPAC Name: 2-[(prop-2-en-1-yloxy)methyl]oxirane
Synonyms: allyl glycidyl ether
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: LSWYGACWGAICNM-UHFFFAOYNA-N
SMILES: C=CCOCC1CO1
Boiling Point 
  • (2)
  • (4)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (6)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (9)
  • (2)
  • (4)
  • (5)
  • (2)
  • (7)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (7)