accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrBr

CAS RN 106-93-4

IUPAC Name: 1,2-dibromoethane
Synonyms: dibromoethane bromofume dwubromoetan caswell no. 439 ethylene dibromides 1,2-dibromoetano soilbrom-90EC E-D-bee unifume DBE
Molecular Weight (g/mol): 187.86
Molecular Formula: C2H4Br2
InChi Key: PAAZPARNPHGIKF-UHFFFAOYSA-N
SMILES: BrCCBr
Molecular Weight (g/mol) 
  • (9)
  • (5)
Boiling Point 
  • (8)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (9)
  • (5)
  • (8)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)