accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 106-94-5

IUPAC Name: 1-bromopropane
Synonyms: 1-bromopropane
Molecular Weight (g/mol): 122.99
Molecular Formula: C3H7Br
InChi Key: CYNYIHKIEHGYOZ-UHFFFAOYSA-N
SMILES: CCCBr
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (5)
  • (5)
  • (2)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (5)