accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-97-8

IUPAC Name: butane
Synonyms: butane pure butane HC 600 (hydrocarbon) LPG N-butane butani A 21 (lowing agent) 0,1 % w/w butadiene diethyl butyl group
Molecular Weight (g/mol): 58.12
Molecular Formula: C4H10
InChi Key: IJDNQMDRQITEOD-UHFFFAOYSA-N
SMILES: CCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Boiling Point 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)
Technique 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results