accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-98-9

IUPAC Name: but-1-ene
Synonyms: 1-butene 1-butene (substance) butene (mixture of isomers)
Molecular Weight (g/mol): 56.11
Molecular Formula: C4H8
InChi Key: VXNZUUAINFGPBY-UHFFFAOYSA-N
SMILES: CCC=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results