accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10603-92-6

IUPAC Name: ethenyltrimethylazanium bromide
Synonyms: ethenyltrimethylazanium bromide
Molecular Weight (g/mol): 166.06
Molecular Formula: C5H12BrN
InChi Key: IDVSELVVGYIOEX-UHFFFAOYSA-M
SMILES: [Br-].C[N+](C)(C)C=C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results