accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10604-21-4

IUPAC Name: 3-phenylcinnoline-4-carboxylic acid
Synonyms: 3-phenylcinnoline-4-carboxylic acid
Molecular Weight (g/mol): 250.26
Molecular Formula: C15H10N2O2
InChi Key: UGJHDXUWDLKCDG-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=NN=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results