CAS RN 10605-21-7
CAS RN 10605-21-7
IUPAC Name:
methyl N-(1H-1,3-benzodiazol-2-yl)carbamate
Synonyms:
carbendazim
Molecular Weight (g/mol):
191.19
Molecular Formula:
C9H9N3O2
InChi Key:
TWFZGCMQGLPBSX-UHFFFAOYSA-N
SMILES:
COC(=O)NC1=NC2=CC=CC=C2N1
1
–
1
of
1
results
1
Sigma Aldrich Carbendazim
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.