accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3ClCl

CAS RN 10605-40-0

IUPAC Name: chloro(3-chloropropyl)dimethylsilane
Synonyms: chloro(3-chloropropyl)dimethylsilane
Molecular Weight (g/mol): 171.14
Molecular Formula: C5H12Cl2Si
InChi Key: BJLJNLUARMMMLW-UHFFFAOYSA-N
SMILES: C[Si](C)(Cl)CCCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results