accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOBrOH

CAS RN 106291-80-9

IUPAC Name: methyl 4-bromo-3-hydroxybenzoate
Synonyms: methyl 4-bromo-3-hydroxybenzoate
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: VYOFPLOREOHCDP-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results