accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106294-60-4

CNCNNNFPFFFFFIr

CAS RN 106294-60-4

IUPAC Name: iridium(3+) 2,2'-bipyridine bis(2-(pyridin-2-yl)benzen-1-ide) hexafluoro-λ⁵-phosphanuide
Synonyms: iridium(3+) bis(2-(pyridin-2-yl)benzen-1-ide) bipyridyl hexafluorophosphate
Molecular Weight (g/mol): 801.75
Molecular Formula: C32H24F6IrN4P
InChi Key: RJJGJTKSOSSNNL-UHFFFAOYSA-N
SMILES: [Ir+3].F[P-](F)(F)(F)(F)F.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)