accessibility menu, dialog, popup

UserName

Viewing profile as user:
FSFFFFF

CAS RN 1063625-86-4

IUPAC Name: 1-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Synonyms: 1-fluoro-4-(pentafluoro-λ⁶-sulfanyl)benzene
Molecular Weight (g/mol): 222.15
Molecular Formula: C6H4F6S
InChi Key: MURGBRJOSPNBEL-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results