accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO(R)O(S)O(S)OHFFF(Z)

CAS RN 106393-80-0

IUPAC Name: (5Z)-7-[(2S,4S,5R)-4-(2-hydroxyphenyl)-2-(trifluoromethyl)-1,3-dioxan-5-yl]hept-5-enoic acid
Synonyms: (5Z)-7-[(2S,4S,5R)-4-(2-hydroxyphenyl)-2-(trifluoromethyl)-1,3-dioxan-5-yl]hept-5-enoic acid
Molecular Weight (g/mol): 374.36
Molecular Formula: C18H21F3O5
InChi Key: ZWAVGFSZMACJHA-PMNBYGLBSA-N
SMILES: OC(=O)CCC\C=C/C[C@@H]1CO[C@@H](O[C@@H]1C1=CC=CC=C1O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results