accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1064-10-4

IUPAC Name: hexaphenyldistannane
Synonyms: hexaphenyldistannane
Molecular Weight (g/mol): 700.06
Molecular Formula: C36H30Sn2
InChi Key: UAPQJVJCJITJET-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)