accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CBr

CAS RN 106418-67-1

IUPAC Name: 1-bromo-4-decylbenzene
Synonyms: 1-bromo-4-decylbenzene
Molecular Weight (g/mol): 297.28
Molecular Formula: C16H25Br
InChi Key: SWTGVSCSSDYINB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (2)