accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOHNCH3

CAS RN 1065073-46-2

IUPAC Name: methyl[(2-methyl-1,3-oxazol-4-yl)methyl]amine
Synonyms: methyl[(2-methyl-1,3-oxazol-4-yl)methyl]amine
Molecular Weight (g/mol): 126.16
Molecular Formula: C6H10N2O
InChi Key: SFLPQWGHHRKLNJ-UHFFFAOYSA-N
SMILES: CNCC1=COC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results