accessibility menu, dialog, popup

UserName

Viewing profile as user:
LiH3CSiCH3CH3O

CAS RN 1066-40-6

IUPAC Name: lithium(1+) trimethylsilanolate
Synonyms: trimethylsilanol
Molecular Weight (g/mol): 96.13
Molecular Formula: C3H9LiOSi
InChi Key: OXOZHAWWRPCVGL-UHFFFAOYSA-N
SMILES: [Li+].C[Si](C)(C)[O-]
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)