accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3CH3HC

CAS RN 1066-54-2

IUPAC Name: ethynyltrimethylsilane
Synonyms: trimethylsilylacetylene
Molecular Weight (g/mol): 98.22
Molecular Formula: C5H10Si
InChi Key: CWMFRHBXRUITQE-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#C
Percent Purity 
  • (3)
  • (10)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (6)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (3)
  • (6)
  • (3)
  • (10)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (6)
  • (4)
  • (6)