accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1066-77-9

SnNHCH3H3CNHCH3CH3NHCH3H3CNHCH3H3C

CAS RN 1066-77-9

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 299.05
Molecular Formula: C8H28N4Sn
InChi Key: YYOYZHVCFBPYPL-UHFFFAOYSA-N
SMILES: [Sn].CNC.CNC.CNC.CNC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results