accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1066-97-3

IUPAC Name: ammonium O,O-dimethyl sulfanidylphosphonothioate
Synonyms: ammonium O,O-dimethyl sulfanidylphosphonothioate
Molecular Weight (g/mol): 175.2
Molecular Formula: C2H10NO2PS2
InChi Key: PPGORMGERPBFTJ-UHFFFAOYSA-N
SMILES: [NH4+].COP([S-])(=S)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results