accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106675-70-1

CH3NOONCH3OH3COCH3

CAS RN 106675-70-1

IUPAC Name: N,N'-dimethoxy-N,N'-dimethylethanediamide
Synonyms: N,N'-dimethoxy-N,N'-dimethylethanediamide
Molecular Weight (g/mol): 176.17
Molecular Formula: C6H12N2O4
InChi Key: OXSWGKZLVLVHIS-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C(=O)N(C)OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)