accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3PH3CCH3CH3OHOCH3OHORu(?)

CAS RN 106681-15-6

IUPAC Name: bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium
Synonyms: bis(acetic acid) {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane ruthenium
Molecular Weight (g/mol): 899.97
Molecular Formula: C52H48O4P2Ru
InChi Key: OXESSJMVRGFBNX-UHFFFAOYSA-N
SMILES: [Ru].CC(O)=O.CC(O)=O.CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)