CAS RN 1067-08-9
IUPAC Name:
3-ethyl-3-methylpentane
Synonyms:
3-methyl-3-ethylpentane
Molecular Weight (g/mol):
114.23
Molecular Formula:
C8H18
InChi Key:
GIEZWIDCIFCQPS-UHFFFAOYSA-N
SMILES:
CCC(C)(CC)CC
1
–
1
of
1
results
1
3-Ethyl-3-methylpentane 99.0+%, TCI America™
CAS: 1067-08-9 Molecular Formula: C8H18 Molecular Weight (g/mol): 114.232 MDL Number: MFCD00048630 InChI Key: GIEZWIDCIFCQPS-UHFFFAOYSA-N PubChem CID: 14018 IUPAC Name: 3-ethyl-3-methylpentane SMILES: CCC(C)(CC)CC