accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClClCl

CAS RN 1067-09-0

IUPAC Name: 1,3-dichloro-2-(chloromethyl)-2-methylpropane
Synonyms: 1,3-dichloro-2-(chloromethyl)-2-methylpropane
Molecular Weight (g/mol): 175.48
Molecular Formula: C5H9Cl3
InChi Key: BYXOMFFBGDPXHB-UHFFFAOYSA-N
SMILES: CC(CCl)(CCl)CCl
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results