accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3COHOOOH

CAS RN 106797-53-9

IUPAC Name: 2-hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methylpropan-1-one
Synonyms: 2-hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methylpropan-1-one
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: GJKGAPPUXSSCFI-UHFFFAOYSA-N
SMILES: CC(C)(O)C(=O)C1=CC=C(OCCO)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)