accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1068-21-9

IUPAC Name: (diethoxyphosphoryl)amine
Synonyms: (diethoxyphosphoryl)amine
Molecular Weight (g/mol): 153.12
Molecular Formula: C4H12NO3P
InChi Key: MCQILDHFZKTBOD-UHFFFAOYSA-N
SMILES: CCOP(N)(=O)OCC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)