accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOOBr

CAS RN 1068-52-6

IUPAC Name: sodium 2-bromoacetate
Synonyms: sodium bromoacetate
Molecular Weight (g/mol): 160.93
Molecular Formula: C2H2BrNaO2
InChi Key: SESSOVUNEZQNBV-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results