accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3MgCl

CAS RN 1068-55-9

IUPAC Name: chloro(propan-2-yl)magnesium
Synonyms: isopropylmagnesium chloride
Molecular Weight (g/mol): 102.84
Molecular Formula: C3H7ClMg
InChi Key: GPIBKUJXKCEZOH-UHFFFAOYSA-M
SMILES: CC(C)[Mg]Cl
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (3)
  • (3)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)