accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3SiCH3CH3H3CSiCH3CH3H3C

CAS RN 1068-69-5

IUPAC Name: [bis(trimethylsilyl)methyl]trimethylsilane
Synonyms: [bis(trimethylsilyl)methyl]trimethylsilane
Molecular Weight (g/mol): 232.59
Molecular Formula: C10H28Si3
InChi Key: BNZSPXKCIAAEJK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C([Si](C)(C)C)[Si](C)(C)C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)