accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1068-90-2

IUPAC Name: 1,3-diethyl 2-acetamidopropanedioate
Synonyms: 1,3-diethyl 2-acetamidopropanedioate
Molecular Weight (g/mol): 217.22
Molecular Formula: C9H15NO5
InChi Key: ISOLMABRZPQKOV-UHFFFAOYSA-N
SMILES: CCOC(=O)C(NC(C)=O)C(=O)OCC
Molecular Weight (g/mol) 
  • (5)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
Quantity 
  • (4)
  • (3)
  • (4)
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (6)
  • (5)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (5)
  • (3)
  • (6)