accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106850-34-4

IUPAC Name: imidazo[1,2-a]pyridine-6-carbonitrile
Synonyms: imidazo[1,2-a]pyridine-6-carbonitrile
Molecular Weight (g/mol): 143.15
Molecular Formula: C8H5N3
InChi Key: LRJOKNYELSECDZ-UHFFFAOYSA-N
SMILES: N#CC1=CN2C=CN=C2C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)