accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHOH

CAS RN 106868-09-1

IUPAC Name: 2,2-dioctylpropane-1,3-diol
Synonyms: 2,2-dioctylpropane-1,3-diol
Molecular Weight (g/mol): 300.53
Molecular Formula: C19H40O2
InChi Key: NFPNQEAEXIXGNY-UHFFFAOYSA-N
SMILES: CCCCCCCCC(CO)(CO)CCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results