accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONH2Br

CAS RN 106896-49-5

IUPAC Name: methyl 4-amino-3-bromobenzoate
Synonyms: methyl 4-amino-3-bromobenzoate
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: AIUWAOALZYWQBX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C(Br)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)