accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3PClCl

CAS RN 1069-08-5

IUPAC Name: (dichlorophosphanyl)diethylamine
Synonyms: (dichlorophosphanyl)diethylamine
Molecular Weight (g/mol): 174
Molecular Formula: C4H10Cl2NP
InChi Key: BPEMCEULJQTJMI-UHFFFAOYSA-N
SMILES: CCN(CC)P(Cl)Cl
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)