accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHNHOOCH3CH3H3C

CAS RN 106930-51-2

IUPAC Name: tert-butyl N-(1-hydroxy-4-methylpentan-2-yl)carbamate
Synonyms: tert-butyl N-(1-hydroxy-4-methylpentan-2-yl)carbamate
Molecular Weight (g/mol): 217.31
Molecular Formula: C11H23NO3
InChi Key: LQTMEOSBXTVYRM-UHFFFAOYNA-N
SMILES: CC(C)CC(CO)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results